Learn More
Thermo Scientific™ POROS™ XS RoboColumn™ Pre-packed columns
Description
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XS resin, a 50-micron strong cation-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS XS resin is the first high-capacity, high-resolution resin that allows loading to more than 100 mg/mL capacity in the presence of up to 15 mS/cm NaCl, while delivering unprecedented separation capability. A major improvement on traditional cation exchangers, POROS XS resin sets the standard for cation exchange chromatography.
Features of the resin:
- Dynamic binding capacity leads to reduced column size, smaller footprint, decreased buffer usage, and reduced cycling
- High resolution supports better product yield and improved impurity clearance
- Robust salt tolerance allows improved process flexibility, alignment, and process flow streamlining
- Low back pressure and linear pressure responses help improve scalability
- Rigid polymeric bead with covalent surface chemistry makes for easier handling and packing, and superior physical and chemical stability, resulting in a robust manufacturing process
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifications
Specifications
| Flow Rate | 16 to 1000 cm/hr. |
| Max. Pressure | 8 bar |
| Stationary Phase | Sulphopropyl Functional Group |
| Column Capacity | 200 μL |
| Quantity | 200 μL |
| Diameter (Metric) Inner | 5 mm |
| Product Line | POROS |
| Shipping Condition | Room Temperature |
| Target | Protein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.