Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ XS RoboColumn™ Pre-packed columns

Catalog No. A30716 Shop All Thermo Scientific Products
Change view
Click to view available options
Column Capacity:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Column Capacity
A30716 600 μL
A30715 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30716 Supplier Thermo Scientific™ Supplier No. A30716
Add to Cart
Add to Cart

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XS resin, a 50-micron strong cation-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.

POROS XS resin is the first high-capacity, high-resolution resin that allows loading to more than 100 mg/mL capacity in the presence of up to 15 mS/cm NaCl, while delivering unprecedented separation capability. A major improvement on traditional cation exchangers, POROS XS resin sets the standard for cation exchange chromatography.

Features of the resin:

  • Dynamic binding capacity leads to reduced column size, smaller footprint, decreased buffer usage, and reduced cycling
  • High resolution supports better product yield and improved impurity clearance
  • Robust salt tolerance allows improved process flexibility, alignment, and process flow streamlining
  • Low back pressure and linear pressure responses help improve scalability
  • Rigid polymeric bead with covalent surface chemistry makes for easier handling and packing, and superior physical and chemical stability, resulting in a robust manufacturing process

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Pore Size Not Applicable
Stationary Phase Sulphopropyl Functional Group
Column Capacity 600 μL
Length (Metric) 3 cm
Quantity 1 Strip, 8 Columns
Particle Size 50 μm
For Use With (Equipment) Robotic Fluid Handlers
Type Cation Exchange Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 30°C. Do not freeze.
Product Line POROS
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Shipping Condition Ambient
Target Protein
Column Format Pre-packed Robo Column
Bed Volume 600 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.