Learn More
Neta Scientific Agilent Poroshell 120,EC-C18, 4.6x50mm,2.7um
SDP
Catalog No. NC9067422
Encompass_Preferred
This item is manufactured by Agilent
Agilent InfinityLab Poroshell 120 EC-C18, 4.6 x 50 mm, 2.7 um, analytical LC column - InfinityLab Poroshell 120
Brand  InfinityLab Poroshell 120
UNSPSC Code  41115709
Particle Size  2.7 um
Separation Mode  Reversed Phase
Phase  EC-C18
- BC
Catalog No. NC9067422 Supplier Neta Scientific Supplier No. 699975902
This item is manufactured by Agilent
Agilent InfinityLab Poroshell 120 EC-C18, 4.6 x 50 mm, 2.7 um, analytical LC column - InfinityLab Poroshell 120
Brand  InfinityLab Poroshell 120
UNSPSC Code  41115709
Particle Size  2.7 um
Separation Mode  Reversed Phase
Phase  EC-C18
- BC