Learn More
Agilent Technologies InfinityLab Poroshell 120 EC-C18, 4.6 x 150 mm, 4 µm, analytical LC column.
Catalog No. NC1483109
Encompass
InfinityLab Poroshell 120 EC-C18, 4.6 x 150 mm, 4 µm, analytical LC column.
Catalog No. NC1483109 Supplier Agilent Technologies Supplier No. 693970902
InfinityLab Poroshell 120 EC-C18, 4.6 x 150 mm, 4 µm, analytical LC column.