Learn More
Agilent Technologies InfinityLab Poroshell 120
Supplier:  Agilent Technologies 699975302
Encompass
InfinityLab Poroshell 120 EC-C18, 3.0 x 50 mm, 2.7 um, solvent saver LC column.
Catalog No. NC1648115