Learn More
Agilent Technologies InfinityLab Poroshell 120
Catalog No. NC1420057
Encompass
InfinityLab Poroshell 120 EC-C18, 3.0 x 100 mm, 2.7 um, solvent saver LC column.
Catalog No. NC1420057 Supplier Agilent Technologies Supplier No. 695975302
InfinityLab Poroshell 120 EC-C18, 3.0 x 100 mm, 2.7 um, solvent saver LC column.