Learn More
Agilent Technologies InfinityLab Poroshell 120 EC-C18
Supplier:  Agilent Technologies 695975302T
Encompass
A 3.0x100mm solvent save LC column with a 2.7um particle size.
Catalog No. NC1773571