Learn More
Chemglass Life Sciences Air Aeration Tube with Sparger for 15L Bio Reactors
SDP
Catalog No. 501294467
Encompass
Air Aeration Tube with Sparger for 15L Bio Reactors. The 15 micron, sintered metal tip produces tiny air bubbles for optimum gas distribution. The A dimension listed allows for placement in different ports of the headplate. NOTE: Replacement O-Ring listed is for M10 connection.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bio reactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4467 Supplier Chemglass Life Sciences Supplier No. CLS138511
Air Aeration Tube with Sparger for 15L Bio Reactors. The 15 micron, sintered metal tip produces tiny air bubbles for optimum gas distribution. The A dimension listed allows for placement in different ports of the headplate. NOTE: Replacement O-Ring listed is for M10 connection.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bio reactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.