Learn More
Chemglass Life Sciences Air Aeration Tube with Sparger for 1L & 2L Bio Reactors
SDP
Catalog No. 501294464
Encompass
Air Aeration Tube with Sparger for 2L Bio Reactors. The 15 micron, sintered metal tip produces tiny air bubbles for optimum gas distribution. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bio reactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4464 Supplier Chemglass Life Sciences Supplier No. CLS137904
Air Aeration Tube with Sparger for 2L Bio Reactors. The 15 micron, sintered metal tip produces tiny air bubbles for optimum gas distribution. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bio reactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.