Learn More
Chemglass Life Sciences Air Aeration Tube with Sparger for 3L Bio Reactors
SDP
Supplier:  Chemglass Life Sciences CLS138012
Encompass
Air Aeration Tube with Sparger for 3L Bio Reactors. The 15 micron, sintered metal tip produces tiny air bubbles for optimum gas distribution. The A dimension listed allows for placement in different ports of the headplate. NOTE: Replacement O-Ring listed is for M10 connection.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bio reactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4552