Learn More
Chemglass Life Sciences Chemglass Air aeration tube with porous sparger for 7 L bioreactors (M10 connection, 330 mm immersion)
SDP
Supplier:  Chemglass Life Sciences CLS138306
Encompass
This air aeration tube with porous sparger is designed for 7 L bioreactors to provide fine gas dispersion through a sintered metal frit. It features stainless steel construction and dimensions suitable for placement in bioreactor headplates.
  • Sintered metal porous tip, 15 μm, for fine bubble generation.
  • 316L stainless steel construction with electropolished finish (20 Ra or better).
  • Immersion length 330 mm; "A" dimension 73 mm for headplate placement.
  • M10 connection for use in compatible headplate ports.
  • Uses replaceable o-ring for sealing.
Catalog No. NC1937660