Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porphobilinogen Pink crystals. MP Biomedicals

Catalog No. ICN15633601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15633601 1 mg
ICN15633605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15633601 Supplier MP Biomedicals, Inc Supplier No. 0215633601
Add to Cart
Add to Cart

Chemical Identifiers

CAS 487-90-1
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.232
InChI Key QSHWIQZFGQKFMA-UHFFFAOYSA-N
Synonym porphobilinogen, unii-74khc72qxk, 3-5-aminomethyl-4-carboxymethyl-1h-pyrrol-3-yl propanoic acid, 1h-pyrrole-3-propanoic acid, 5-aminomethyl-4-carboxymethyl, 2-aminomethylpyrrol-3-acetic acid 4-propionic acid, 74khc72qxk, pbg, 5-aminomethyl-4-carboxymethyl-1h-pyrrole-3-propanoic acid, 5-aminomethyl-4-carboxymethyl-1h-pyrrole-3-propionic acid, 5-aminomethyl-4-carboxymethyl-pyrrole-3-propionic acid
PubChem CID 1021
ChEBI CHEBI:17381
IUPAC Name 3-[5-(aminomethyl)-4-(carboxymethyl)-1H-pyrrol-3-yl]propanoic acid
SMILES C1=C(C(=C(N1)CN)CC(=O)O)CCC(=O)O

Specifications

Color Pink
Quantity 1 mg
Formula Weight 226.2
Physical Form Crystals
Chemical Name or Material Porphobilinogen
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.