Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5mg, Porphobilinogen, >97%, [487-90-1], P226 244.24 C10H14N2O4 x H2O

Catalog No. 501455901
Encompass_Preferred

Compound: Porphobilinogen

CAS#: 487-90-1

MDL#: not listed

Background: A pyrrole-based natural product that serves as a starting material for the biosynthesis of porphyrins such as heme and chlorophyll. It is also a main indicator of altered metabolism, indicative of lead poisoning, and a biomarker for porphyria disease.

Storage: Store at room temperature, protected from light.

Catalog No. 50-145-5901 Supplier Frontier Specialty Chemicals Supplier No. P2265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Porphobilinogen

CAS#: 487-90-1

MDL#: not listed

Background: A pyrrole-based natural product that serves as a starting material for the biosynthesis of porphyrins such as heme and chlorophyll. It is also a main indicator of altered metabolism, indicative of lead poisoning, and a biomarker for porphyria disease.

Storage: Store at room temperature, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.