Learn More
ATCC Porphyromonas gingivalis; 2561
Catalog No. 502381920
Encompass
Porphyromonas gingivalis strain 2561 is a whole-genome sequenced bacterial type strain that was isolated from a human gingival sulcus. This culture is part of a set of quality control organisms for BioMerieux Vitek products.
Catalog No. 50-238-1920 Supplier ATCC Supplier No. 33277
Porphyromonas gingivalis strain 2561 is a whole-genome sequenced bacterial type strain that was isolated from a human gingival sulcus. This culture is part of a set of quality control organisms for BioMerieux Vitek products.