Learn More
Stoelting™ Porsolt Forced Swim Test
SDP
Catalog No. 10000304 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
10-000-304 Mouse, Rat
1 options
Catalog No. 10-000-304 Supplier Stoelting™ Supplier No. 60160
Add to Cart
Add to Cart
Stoelting™ Porsolt Forced Swim Test is used to test for assessment of depression in animal models.
Measures the effect of antidepressant drugs on behavior.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat