Learn More
Cole-Parmer Idex P-311 Port Plug, Standard Knurl, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA
Catalog No. NC1748532 Shop All Cole-Parmer Products
Encompass
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
Catalog No. NC1748532 Supplier Cole-Parmer Supplier No. 0201378
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.