Learn More
Azzota Corp Portable Salinity Refractometer, Salinity Range: 0-28%, Resolution: 0.2%, Accuracy +/-0.2%, With Atc
Catalog No. 501612922
Encompass_Preferred
Portable Salinity Refractometer, Salinity range: 0-28%, Resolution: 0.2%, Accuracy +/-0.2%, with ATC
Catalog No. 50-161-2922 Supplier Azzota Corp Supplier No. REF28T
Portable Salinity Refractometer, Salinity range: 0-28%, Resolution: 0.2%, Accuracy +/-0.2%, with ATC