Learn More
Cole-Parmer Dwyer A-432 Portability Kit for Magnehelic Gauges
Catalog No. 50992875 Shop All Cole-Parmer Products
Encompass_Preferred
Plastic carrying case for Magnehelic gage of standard range, except high pressure connection, includes 9 ft (2.7m) of 3/16" ID rubber tubing, stand-hang bracket & terminal tube with holder.
Catalog No. 50-992-875 Supplier Cole-Parmer Supplier No. 6846277
Plastic carrying case for Magnehelic gage of standard range, except high pressure connection, includes 9 ft (2.7m) of 3/16" ID rubber tubing, stand-hang bracket & terminal tube with holder.