Learn More
Sentry Air Systems Model 300 Portable Room Air Cleaner
SDP
Catalog No. NC2885520
Encompass
The Portable Room Air Cleaner combines efficient air filtration and compact design for an air quality improvement solution. This system features heavy-duty casters and a variable speed control. The Portable Room Air Cleaner offers a quiet low-power solution for general air filtration in medical and laboratory environments but also can be used as a secondary control solution for fume and particulates. This system works by drawing fumes and particulate into the filtration system and releases the filtered air back into the surrounding room creating a recirculating airflow. Multiple filters can be used to control both fumes and particulate. Approximate Airflow: up to 350 CFM
Catalog No. NC2885520 Supplier Sentry Air Systems Supplier No. SS300PRAC
The Portable Room Air Cleaner combines efficient air filtration and compact design for an air quality improvement solution. This system features heavy-duty casters and a variable speed control. The Portable Room Air Cleaner offers a quiet low-power solution for general air filtration in medical and laboratory environments but also can be used as a secondary control solution for fume and particulates. This system works by drawing fumes and particulate into the filtration system and releases the filtered air back into the surrounding room creating a recirculating airflow. Multiple filters can be used to control both fumes and particulate. Approximate Airflow: up to 350 CFM