Learn More
Electron Microscopy Sciences Portable Air Compressor
SDP
Catalog No. 5033087
Encompass_Preferred
Pressurize the Sure Shot Sprayers with this rechargeable battery powered Air Compressor. The Compressor generates up to 220 PSI, the battery recharge unit by the AC 115V adapter or DC 12V adapter included. Completely portable, no trailing cords.
Catalog No. 50-330-87 Supplier Electron Microscopy Sciences Supplier No. 64195
Pressurize the Sure Shot Sprayers with this rechargeable battery powered Air Compressor. The Compressor generates up to 220 PSI, the battery recharge unit by the AC 115V adapter or DC 12V adapter included. Completely portable, no trailing cords.