Learn More
Grainger Portable Band Saw Kit 6.5A
Catalog No. 502751994
Encompass_Preferred
The Makita Portable Band Saw Kit includes a 6.5 Amp motor band saw that can cut up to 4-3/4" round stock. It has variable speed control from 260-530 FPM. The kit comes with a tool bag, hex wrench, blade, and blade guide rollers. It weighs 15 lbs and has a built-in LED light. The saw can make cuts up to 5" deep and has a cutting capacity of 4-3/4" round stock. It is designed for one-handed operation.
Catalog No. 50-275-1994 Supplier Grainger Supplier No. 14U140
The Makita Portable Band Saw Kit includes a 6.5 Amp motor band saw that can cut up to 4-3/4" round stock. It has variable speed control from 260-530 FPM. The kit comes with a tool bag, hex wrench, blade, and blade guide rollers. It weighs 15 lbs and has a built-in LED light. The saw can make cuts up to 5" deep and has a cutting capacity of 4-3/4" round stock. It is designed for one-handed operation.