Learn More
Azzota Corp Portable Brix refract 0-20pct
Catalog No. 5000887289
Encompass_Preferred
Heavy-Duty Portable Refractometer Sturdy design made of Aluminum Brix Range 0-20% Brix Min Div 1% Brix Accuracy /-1 0% With ATC
Catalog No. 50-008-87289 Supplier Azzota Corp Supplier No. RHB20T
Heavy-Duty Portable Refractometer Sturdy design made of Aluminum Brix Range 0-20% Brix Min Div 1% Brix Accuracy /-1 0% With ATC