Learn More
Grainger Vestil Cylinder Lifting Dolly, Steel, 300 lb. Capacity, 1 ea
Catalog No. 502776237
Encompass_Preferred
This portable cylinder lifter is designed for the safe and efficient movement and lifting of gas cylinders for placement into portable welders. Featuring a durable steel construction with a yellow powder-coated finish, this unit accommodates cylinders with a maximum diameter of 11.5 in. and a maximum height of 56 in. The cylinder is secured to the cart using a manual ratchet strap, while a manual hand crank cable winch facilitates vertical movement. The unit is equipped with two 10 in. hard rubber wheels for portability.
  • 300 lb. uniform weight capacity
  • Manual hand crank cable winch for controlled lifting
  • Manual ratchet strap for secure cylinder attachment
  • Steel construction with a protective yellow powder-coated finish
  • Hard rubber wheels for smooth mobility
  • Designed for use in industrial applications
Catalog No. 50-277-6237 Supplier Grainger Supplier No. 172N48
This portable cylinder lifter is designed for the safe and efficient movement and lifting of gas cylinders for placement into portable welders. Featuring a durable steel construction with a yellow powder-coated finish, this unit accommodates cylinders with a maximum diameter of 11.5 in. and a maximum height of 56 in. The cylinder is secured to the cart using a manual ratchet strap, while a manual hand crank cable winch facilitates vertical movement. The unit is equipped with two 10 in. hard rubber wheels for portability.
  • 300 lb. uniform weight capacity
  • Manual hand crank cable winch for controlled lifting
  • Manual ratchet strap for secure cylinder attachment
  • Steel construction with a protective yellow powder-coated finish
  • Hard rubber wheels for smooth mobility
  • Designed for use in industrial applications