Learn More
MSC Multi-Clean U-Fill Foamer Gun, Portable, 25-85 PSI
Catalog No. 502837575
Encompass
This portable foaming unit produces thick, clinging foam for enhanced cleaning performance on vertical surfaces. Ideal for restrooms, showers, and heavy soil areas, it helps increase chemical dwell time while reducing waste and labor.
  • Allows dispensing of foaming cleaners directly onto vertical surfaces.
  • Features an on/off trigger valve for easy control.
  • Includes a quick disconnect for convenience.
  • Rotating selector valve allows switching between product application and water rinse.
  • Requires a tap water supply with pressure between 25 and 85 PSI.
  • Increases chemical dwell time on surfaces for improved effectiveness.
  • Reduces chemical waste and labor.
  • Ideal for removing soap scum and mineral deposits.
Catalog No. 50-283-7575 Supplier MSC Supplier No. 43473024
This portable foaming unit produces thick, clinging foam for enhanced cleaning performance on vertical surfaces. Ideal for restrooms, showers, and heavy soil areas, it helps increase chemical dwell time while reducing waste and labor.
  • Allows dispensing of foaming cleaners directly onto vertical surfaces.
  • Features an on/off trigger valve for easy control.
  • Includes a quick disconnect for convenience.
  • Rotating selector valve allows switching between product application and water rinse.
  • Requires a tap water supply with pressure between 25 and 85 PSI.
  • Increases chemical dwell time on surfaces for improved effectiveness.
  • Reduces chemical waste and labor.
  • Ideal for removing soap scum and mineral deposits.