Learn More
Grainger Portable Gas End Diffuser 19 1/2 H
Catalog No. 502765617
Encompass_Preferred
The L.B. White Portable Gas Heater Diffuser is a portable gas-fired construction heater that provides up to 250,000 BTU/hr of heat output. It features a durable steel housing, integrated carrying handles, and an adjustable heat diffuser to direct heat where needed. This portable construction heater runs on propane or natural gas, has a rust resistant finish, and includes a 10 ft hose with regulator and quick disconnects for easy hookup. With an automatic ignition and thermostat control, this portable heater provides temporary heating for construction sites, warehouses, garages, and other applications.
Catalog No. 50-276-5617 Supplier Grainger Supplier No. 16W313
The L.B. White Portable Gas Heater Diffuser is a portable gas-fired construction heater that provides up to 250,000 BTU/hr of heat output. It features a durable steel housing, integrated carrying handles, and an adjustable heat diffuser to direct heat where needed. This portable construction heater runs on propane or natural gas, has a rust resistant finish, and includes a 10 ft hose with regulator and quick disconnects for easy hookup. With an automatic ignition and thermostat control, this portable heater provides temporary heating for construction sites, warehouses, garages, and other applications.