Learn More
Grainger Portable Gas Torpedo HeatrLP 1050 cfm
Supplier:  Grainger 16W282
Encompass_Preferred
The L.B. White Portable Gas Torpedo Heater is a portable propane heater that provides up to 100,000 BTUs of heat output. It features a fully enclosed, weather-resistant steel cabinet with a powder coat finish and four swivel casters for easy mobility. This torpedo heater has an electronic ignition system for reliable startups and can heat areas up to 2,500 square feet. Safety features include a tip-over switch that automatically shuts off the heater if tilted more than 15 degrees. This portable propane heater requires no electricity to operate and includes a 10-foot hose and regulator for connecting to a propane tank (sold separately).
Catalog No. 50-276-6195