Learn More
Grainger L.B. White Premier® 80 2.0 Portable Heater, Steel, LP Gas, 1 ea
Catalog No. 502767064
Encompass_Preferred
The Premier® 80 2.0 is an enclosed flame, direct-fired portable heater designed to provide dependable heating and air circulation for tents and temporary spaces. Featuring an efficient design constructed from durable steel, this heater is built to withstand extreme outdoor environments while delivering high-volume air output.
  • 99% fuel-efficient operation
  • Totally enclosed flame design for safety
  • Operates on LP gas
  • Reliable electronic spark ignition
  • CSA U.S., CSA Canada, and CE Type certified
  • Includes hose, regulator, and thermostat
  • Equipped with lift handles and semi-pneumatic wheels for easy transport
  • Suitable for indoor or outdoor placement
Catalog No. 50-276-7064 Supplier Grainger Supplier No. 16W305
The Premier® 80 2.0 is an enclosed flame, direct-fired portable heater designed to provide dependable heating and air circulation for tents and temporary spaces. Featuring an efficient design constructed from durable steel, this heater is built to withstand extreme outdoor environments while delivering high-volume air output.
  • 99% fuel-efficient operation
  • Totally enclosed flame design for safety
  • Operates on LP gas
  • Reliable electronic spark ignition
  • CSA U.S., CSA Canada, and CE Type certified
  • Includes hose, regulator, and thermostat
  • Equipped with lift handles and semi-pneumatic wheels for easy transport
  • Suitable for indoor or outdoor placement