Learn More
Electron Microscopy Sciences Portable Heat Pen
SDP
Catalog No. 50949523
Encompass_Preferred
This portable heat pen is able to heat up to 700 degree C and automatically recharges every time it is placed back into its recharging stand. It will last approximately 40 minutes on a full charge. Tip length 1". It is useful for the sealing of wax onto glass knife boats and small soldering jobs around the lab. The set includes the heat pen and recharging stand.
Catalog No. 50-949-523 Supplier Electron Microscopy Sciences Supplier No. 7267710
This portable heat pen is able to heat up to 700 degree C and automatically recharges every time it is placed back into its recharging stand. It will last approximately 40 minutes on a full charge. Tip length 1". It is useful for the sealing of wax onto glass knife boats and small soldering jobs around the lab. The set includes the heat pen and recharging stand.