Learn More
HANNA INSTRUMENTS INC Portable Lux Meter
SDP
Catalog No. NC1749685
Encompass
HI97500 is a portable lux meter designed for simplicity of use in taking rapid light measurements. The HI97500 comprises a rugged, water-resistant housing of the meter and durable casing of the light sensor. Additional features include three measurement ranges, an auto-off function after 7 minutes of inactivity, and low battery indicator.Three measurement rangesWater-resistant housing200 hour battery life with battery level indicator
Catalog No. NC1749685 Supplier HANNA INSTRUMENTS INC Supplier No. HI97500
HI97500 is a portable lux meter designed for simplicity of use in taking rapid light measurements. The HI97500 comprises a rugged, water-resistant housing of the meter and durable casing of the light sensor. Additional features include three measurement ranges, an auto-off function after 7 minutes of inactivity, and low battery indicator.Three measurement rangesWater-resistant housing200 hour battery life with battery level indicator