Learn More
Azzota Corp Portable Ph/Mv/Orp/ C/ F Meter, Resolution: 0.01 Ph / 1 Mv / 0.1'C, Ph Range: -2.00~16.00Ph, Mv Range: 0 To +/-1999.9Mv, Temperature Range: 0 To 100.0'C (32 To 212'F)
Catalog No. 501612907
Encompass_Preferred
Portable pH/mV/ORP/ C/ F Meter, Resolution: 0.01 pH / 1 mV / 0.1'C, pH Range: -2.00~16.00pH, mV Range: 0 to +/-1999.9mV, Temperature Range: 0 to 100.0'C (32 to 212'F)
Catalog No. 50-161-2907 Supplier Azzota Corp Supplier No. PHMVT220
Portable pH/mV/ORP/ C/ F Meter, Resolution: 0.01 pH / 1 mV / 0.1'C, pH Range: -2.00~16.00pH, mV Range: 0 to +/-1999.9mV, Temperature Range: 0 to 100.0'C (32 to 212'F)