Learn More
Bellco Glass, Inc. Portable Pip-Aid,115V
SDP
Catalog No. 501944624
Encompass_Preferred
The original portable pipet-aid is rechargeable self contained unit that provides 2 hours of pipetting before recharging is necessary. Includes battery charger and tissue culture nose piece. Pipets over 2000 times without recharging
Catalog No. 50-194-4624 Supplier Bellco Glass, Inc. Supplier No. 122585000
The original portable pipet-aid is rechargeable self contained unit that provides 2 hours of pipetting before recharging is necessary. Includes battery charger and tissue culture nose piece. Pipets over 2000 times without recharging