Learn More
USA Scientific Inc Portable Pipet-Aid XL
Catalog No. 507541140
Encompass
Portable Pipet-Aid XL battery operated controller with long handle. Includes wall hanger, 110V charger, and four replacement filters.
Catalog No. 50-754-1140 Supplier USA Scientific Inc Supplier No. 44400105
Portable Pipet-Aid XL battery operated controller with long handle. Includes wall hanger, 110V charger, and four replacement filters.