Learn More
Bellco Glass, Inc. Portable Pipet-Aid XPモ,220V-CE W/Charger & 4 Repl Filters
SDP
Catalog No. 501944627
Encompass_Preferred
Specifically designed to minimize operator fatigue, the portable pipet-aid xp is smaller and lightweight. It accepts most pipets from 1-100ml. Each button has three speed settings ( fast, medium, slow). Medium speed is suitable for most routine techniques
Catalog No. 50-194-4627 Supplier Bellco Glass, Inc. Supplier No. 122586220
Specifically designed to minimize operator fatigue, the portable pipet-aid xp is smaller and lightweight. It accepts most pipets from 1-100ml. Each button has three speed settings ( fast, medium, slow). Medium speed is suitable for most routine techniques