Learn More
USA Scientific Inc Portable Pipet-AidxP2, 1 unit
Catalog No. 502745046
Encompass
Portable Pipet-Aid XP2 battery operated controller for serological pipets, includes recharger, bench stand, and filters
Catalog No. 50-274-5046 Supplier USA Scientific Inc Supplier No. 44405010
Portable Pipet-Aid XP2 battery operated controller for serological pipets, includes recharger, bench stand, and filters