Description
- Patented lock rails hold up to 16 padlocks and the slot in front allows keys to be inserted when box is locked
- Use one lock on a each energy control point and place the keys in the lockout box
- each worker then places his own lock on the box to prevent access
- Each employee retains exclusive control, as required by OSHA, by placing his own lock on the lockout box containing the keys to the job locks
- As long as any one workers lock remains on the lockout box, the keys to the job locks contained inside cannot be accessed
Specifications
Specifications
| Product Type | Lock Box |
| Color | Red |
| Quantity | 1 Ea. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.