Learn More
3M™ Portable Radio Push-to-Talk (PTT) Adaptors
Catalog No. 19065967 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Kenwood TK220/320, 240/340, 430/431, 250/350/353,260/360, 270/370, Free Talk, 3100 Connection Headsets
Kenwood TK480/481, 290/390/380, TK2140/3140 Connection Headsets
Motorola HT750, HT1250, HT1550, GP328, GP338, MTS-8250, MTX850LS, MTX8250LS, MTX8259LS, MTX950 Connection Headsets
MotorolaVisar, XTS3000, XTS-5000, GP900, HT1000, MT2000, JT1000, MTS2000, MTX838/2000/8000/9000 Connection Headsets
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-065-967 Kenwood TK480/481, 290/390/380, TK2140/3140 Connection Headsets
19-065-958 Motorola HT750, HT1250, HT1550, GP328, GP338, MTS-8250, MTX850LS, MTX8250LS, MTX8259LS, MTX950 Connection Headsets
19-065-957 MotorolaVisar, XTS3000, XTS-5000, GP900, HT1000, MT2000, JT1000, MTS2000, MTX838/2000/8000/9000 Connection Headsets
19-065-959 Kenwood TK220/320, 240/340, 430/431, 250/350/353,260/360, 270/370, Free Talk, 3100 Connection Headsets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-065-967 Supplier 3M™ Supplier No. 7100018196
Add to Cart
Add to Cart
This is an adapter compatible with the 3M™ PELTOR™ Series. It works in conjunction with the Motorola GP300, CP200, Radius P111 communication radios along with various PELTOR™ headsets.
This in-line Push-to-Talk (PTT) adapter is compatible with the 3M™ Peltor™ Solutions and works in conjunction with the Kenwood TK380 communication radio. This adapter works in conjunction with 2-way radio communication set-ups along with various Peltor™ MT Series headsets.
  • Waterproof
  • 360-degree rotating belt clip assembly
  • Easy-to-activate palm button design
  • 3M Catalog# FL5014, FL5018, FL5030, FL5035, FL5078
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push-to-Talk Adaptor
For Use With (Equipment) Kenwood TK480/481, 290/390/380, TK2140/3140 Connection Headsets
Model FL5078