Learn More
Felix Storch Inc ACCUCOLD PORTABLE REFRIG/FREEZER
SDP
Catalog No. NC2874829
Encompass_Preferred
The SPRF11 can be used as a refrigerator or freezer, with a wide temperature range that can be adjusted between -20ºC and +20ºC, ensuring vaccines can be stored at the recommended 2 to 8ºC range. The external control panel offers easy and intuitive operation and allows you to adjust the temperature without opening the lid and exposing stored items to ambient temperature. This unit is also equipped with a heating function that lets you set the interior at +50ºC, a useful feature for warming compression packs. With its user-friendly portable design, the SPRF11 is a great solution for traveling nurses, off-site vaccination clinics, sample collection, environmental agencies, and other applications that require temperature stability while in transit: https://www.accucold.com/product/SPRF11
Catalog No. NC2874829 Supplier Felix Storch Inc Supplier No. SPRF11
The SPRF11 can be used as a refrigerator or freezer, with a wide temperature range that can be adjusted between -20ºC and +20ºC, ensuring vaccines can be stored at the recommended 2 to 8ºC range. The external control panel offers easy and intuitive operation and allows you to adjust the temperature without opening the lid and exposing stored items to ambient temperature. This unit is also equipped with a heating function that lets you set the interior at +50ºC, a useful feature for warming compression packs. With its user-friendly portable design, the SPRF11 is a great solution for traveling nurses, off-site vaccination clinics, sample collection, environmental agencies, and other applications that require temperature stability while in transit: https://www.accucold.com/product/SPRF11