Learn More
Grainger Portable Safe 0.05 cu ft Black
Catalog No. 502766697
Encompass_Preferred
The Grainger 16X139 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade level (from 5 to 13) and delay controls, and comfortable padded headgear. The helmet provides versatility for most welding applications.
Catalog No. 50-276-6697 Supplier Grainger Supplier No. 16X139
The Grainger 16X139 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade level (from 5 to 13) and delay controls, and comfortable padded headgear. The helmet provides versatility for most welding applications.