Learn More
Mopec Portable Side-Loading Cadaver Rack, 32 in.
SDP
Catalog No. 21403188 Shop All Mopec Products
Change view
Click to view available options
Tiers:
3
4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tiers
21-403-188 4
21-403-187 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-188 Supplier Mopec Supplier No. IW32401
Built from durable stainless steel for reliable daily use in laboratory and post-mortem environments.
  • Designed to accommodate 31 in. body trays (not included) with smooth side-loading functionality, featuring ten full-width rollers per tier for controlled and efficient tray movement in laboratory and morgue settings.
  • Equipped with four rubber-bumpered tray stops and positive-locking mechanisms on each tier to enhance safety, stability, and secure positioning during loading and unloading.
  • Manufactured from thick-wall 304 stainless steel with a 180-grit finish, ensuring corrosion resistance, easy cleaning, and suitability for post-mortem and medical environments.
  • Available in two configurations:
    • IW32301 measuring 83.0 L x 34.0 W x 64.7 H in. with tier heights of 23.6, 42.6, and 59.6 in.
    • IW32401 measuring 83.0 L x 34.0 W x 72.6 H in. with tier heights of 23.6, 42.6, 59.6, and 68.6 in.
  • Engineered to support heavy-duty use with a load capacity of 500 lbs per tier, suitable for daily operational demands.
  • Compatible with GA031 31 in. trays, JD5001 scissor lift, and JD450, JD451, M677, and M678 forklifts when used with JD460 reversible forks and JD420 roller pallets.
  • Additional sizes and accessories are available to support varied facility requirements.
Specifications
Product Type Cadaver Storage Rack
Tiers 4