Learn More
Braintree Scientific Portable Small Animal Warming Plate
SDP
Catalog No. 501955588
Encompass
Portable Small Animal Warming Plate Use under animal to prevent hypothermia during surgery, post-surgery recovery and anesthesia induction. Small enough to place under animals during stereotaxic surgery. Temperature range: 31C, 35C, 45C Can be placed on bench or in cage Non-skid feet for added stability HD power supply with universal input for use in any country (100 240 VAC, 60 Hz) 7" x4", stainless steel **Sold for research purposes only and not for human use**
Catalog No. 50-195-5588 Supplier Braintree Scientific Supplier No. PWPV
Portable Small Animal Warming Plate Use under animal to prevent hypothermia during surgery, post-surgery recovery and anesthesia induction. Small enough to place under animals during stereotaxic surgery. Temperature range: 31C, 35C, 45C Can be placed on bench or in cage Non-skid feet for added stability HD power supply with universal input for use in any country (100 240 VAC, 60 Hz) 7" x4", stainless steel **Sold for research purposes only and not for human use**