Learn More
Brady™ Poly-Edge Mountable Spill Kit
Catalog No. 19103130 Shop All Brady Corporation Products
Catalog No. 19-103-130 Supplier Brady™ Supplier No. SKOTRK/PVT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Improve your levels of emergency spill protection and reaction. The Brady™ Poly-Edge Mountable Spill Kit has everything you need to be prepared.
  • The oil only Poly-Edge Spill Kit is lightweight for maximum mobility
  • 100% polyethlyene construction so it will not rust or corrode
  • Lockable container prevents tampering
  • Flexible gasket allows for wall or truck mounting
Specifications
Type Spill Kit, Oil-only
For Use With (Application) Oil-only spills
Includes Versatile Poly-Edge container, (25) 15 in. x 19 in. pads, (4) 3 in. x 12 in. socks, 1 drain cover, 1 pair of nitrile gloves, 1 pair of goggles, 1 hazmat suit, 1 putty pat, 2 disposal bags, 1 instruction sheet
Packaging Quantity 1 Kit