Learn More
MSC Portacool Replacement Pump, 0.2 Amps, for APEX/Cyclone Coolers
Catalog No. 5000509886
Encompass
This replacement pump is a genuine authorized replacement part designed to meet the exact performance requirements of Portacool-branded portable evaporative coolers, including the APEX 500, APEX 700, Cyclone 120, Cyclone 130, and Portacool 510 series.
  • Genuine authorized replacement part.
  • Fits APEX 500 and APEX 700 evaporative coolers.
  • Fits Cyclone 120 and 130 evaporative coolers and 510 models.
  • Compatible with discontinued Cyclone 110, 1000, 2000, 2200, 3000 and 3200 models.
  • Preassembled with quick connect for easy installation.
  • Installation resources are available online and through the customer care team.
Catalog No. 50-005-09886 Supplier MSC Supplier No. 71543458
This replacement pump is a genuine authorized replacement part designed to meet the exact performance requirements of Portacool-branded portable evaporative coolers, including the APEX 500, APEX 700, Cyclone 120, Cyclone 130, and Portacool 510 series.
  • Genuine authorized replacement part.
  • Fits APEX 500 and APEX 700 evaporative coolers.
  • Fits Cyclone 120 and 130 evaporative coolers and 510 models.
  • Compatible with discontinued Cyclone 110, 1000, 2000, 2200, 3000 and 3200 models.
  • Preassembled with quick connect for easy installation.
  • Installation resources are available online and through the customer care team.