Learn More
SAI Infusion Technologies Rat Portal Vein Catheter - 31cm with 1cm tip and 4 suture beads
SDP
Catalog No. NC3455018
Encompass
Portal Vein Catheter for Rats. 31cm PU catheter including a 1cm PE tip and 4 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.
Catalog No. NC3455018 Supplier SAI Infusion Technologies Supplier No. RPCPUX30PE
Portal Vein Catheter for Rats. 31cm PU catheter including a 1cm PE tip and 4 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.