Learn More
Med Vet International Portex Equine Stomach Tube, Foal, 1/4" x 3/8" x 7", J0106F
Supplier:  Med Vet International JOR106F
Encompass_Preferred
This is the finest quality tube on the market. Made from special siliconized PVC tubing, it has just the right amount of flexibility; it won’t become rigid in cold weather and will not soften or kink in hot weather. Each tube has a funnel end to make attachment to a drench pump quick and easy.
Catalog No. 50-005-83761