Learn More
Med Vet International Portex Equine Stomach Tube, Large, 1/2" x 3/4" x 10", J0106L
Catalog No. 5000583488
Encompass_Preferred
This is the finest quality tube on the market. Made from special siliconized PVC tubing, it has just the right amount of flexibility; it won’t become rigid in cold weather and will not soften or kink in hot weather. Each tube has a funnel end to make attachment to a drench pump quick and easy.
Catalog No. 50-005-83488 Supplier Med Vet International Supplier No. JOR106L
This is the finest quality tube on the market. Made from special siliconized PVC tubing, it has just the right amount of flexibility; it won’t become rigid in cold weather and will not soften or kink in hot weather. Each tube has a funnel end to make attachment to a drench pump quick and easy.