Learn More
Waters Corp HIC Protein Standard Mix, 6-component, Lyophilized Solid, Reference
Catalog No. NC0833847 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HIC Protein Standard Mix is a 6-component intact protein mixture for benchmarking HIC columns, aiding in system monitoring, method development, and troubleshooting.
  • Formulated with Ribonuclease A, Cytochrome C, Myoglobin, Lysozyme, Yeast Enolase, and alpha-chymotrypsinogen A.
  • Packaged as a lyophilized solid in a TruView vial.
  • Solubilize in 100 μL distilled water or a 50/50 mix of Mobile Phase A and B; store at -20 °C.
  • Assesses column performance and serves as a reference for accurate protein quantification.
  • Stable formulation ensures consistent, reproducible results and seamless workflow integration.
Catalog No. NC0833847 Supplier Waters Corp Supplier No. 186007953
The HIC Protein Standard Mix is a 6-component intact protein mixture for benchmarking HIC columns, aiding in system monitoring, method development, and troubleshooting.
  • Formulated with Ribonuclease A, Cytochrome C, Myoglobin, Lysozyme, Yeast Enolase, and alpha-chymotrypsinogen A.
  • Packaged as a lyophilized solid in a TruView vial.
  • Solubilize in 100 μL distilled water or a 50/50 mix of Mobile Phase A and B; store at -20 °C.
  • Assesses column performance and serves as a reference for accurate protein quantification.
  • Stable formulation ensures consistent, reproducible results and seamless workflow integration.