Learn More
Dataworks Development Inc PORTRAIT CRYO LABELS-1ML 2.75
SDP
Catalog No. NC9569413
Encompass
This item has a minimum qty of 5 per supplier requirements.
Cryogenic safe wraparound labels. Self-laminating, protects ink from degradation in harsh environments. 1.00 in.x 2.75 in. with printable area of 1.3 in. x 1.00 in.. Use on 1-2ml cryovials for long term storage in all freezers and LN2 tanks.
Catalog No. NC9569413 Supplier Dataworks Development Inc Supplier No. LBL02WR1
This item has a minimum qty of 5 per supplier requirements.
Cryogenic safe wraparound labels. Self-laminating, protects ink from degradation in harsh environments. 1.00 in.x 2.75 in. with printable area of 1.3 in. x 1.00 in.. Use on 1-2ml cryovials for long term storage in all freezers and LN2 tanks.