Learn More
THORLABS INC POS GRID DISTORTION TARGET,3X1
Catalog No. NC2185918
Encompass
Positive Grid Distortion Target, 3" x 1", 10, 50, 100, and 500 µm Grid Spacings
This item is not returnable. View return policy
Catalog No. NC2185918 Supplier THORLABS INC Supplier No. R1L3S3P
Positive Grid Distortion Target, 3" x 1", 10, 50, 100, and 500 µm Grid Spacings