Learn More
JANT PHARMACAL CORPORATION Pregnancy-Skreen hCG Controls. One 4 mL Pos and One 4 mL Neg.
SDP
Catalog No. NC3457004
Encompass_Preferred
The PREGNANCY-SKREEN™ Human Chorionic Gonadotropin (hCG) Positive and Negative controls are prepared from human based urine. These hCG controls can be used to monitor the performance of qualitative immunochromatographic hCG detection devices.
Catalog No. NC3457004 Supplier JANT PHARMACAL CORPORATION Supplier No. PC57
The PREGNANCY-SKREEN™ Human Chorionic Gonadotropin (hCG) Positive and Negative controls are prepared from human based urine. These hCG controls can be used to monitor the performance of qualitative immunochromatographic hCG detection devices.