Learn More
AllCells LLC Peripheral Blood, Cryopreserved, CD56+ Natural Killer (NK) Cells, Positively Selected, 5M
Catalog No. 50156584
Encompass
AllCells Positively Selected NPB-CD56+ NK cells are directly isolated from mononuclear cells using direct immunomagnetic CD56 MicroBeads.
Catalog No. 50-156-584 Supplier AllCells LLC Supplier No. LP, CR, CD56+, PS, 5M
AllCells Positively Selected NPB-CD56+ NK cells are directly isolated from mononuclear cells using direct immunomagnetic CD56 MicroBeads.